2-Amino-N-(2-methylphenyl)-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053321-500MG
053321-2.5G
101903-170EA
364.62
USD
101903-170
101903-172
2-Amino-N-(2-methylphenyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(2-methylphenyl)-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₆H₁₉ClN₂O |
MDL Number:
MFCD13562710 CAS Number: 1236261-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...