3-Amino-N-(2-methylphenyl)propanamide hydrochloride

Supplier: Matrix Scientific

053318-500MG 053318-2.5G
101903-158EA 364.62 USD
101903-158 101903-160
3-Amino-N-(2-methylphenyl)propanamide hydrochloride
3-Amino-N-(2-methylphenyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD13194012
CAS Number: 938515-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR