2-(Methylamino)-N-(2-methylphenyl)acetamide hydrochloride

Supplier: Matrix Scientific

053317-500MG 053317-2.5G
101903-154EA 364.62 USD
101903-154 101903-156
2-(Methylamino)-N-(2-methylphenyl)acetamide hydrochloride
2-(Methylamino)-N-(2-methylphenyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD01670960
CAS Number: 1049701-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR