2-Amino-N-(2-methylphenyl)propanamide hydrochloride

Supplier: Matrix Scientific

053316-500MG 053316-2.5G
101903-150EA 149.23 USD
101903-150 101903-152
2-Amino-N-(2-methylphenyl)propanamide hydrochloride
2-Amino-N-(2-methylphenyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD13191984
CAS Number: 35891-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR