2-Amino-N-(2-methylphenyl)acetamide hydrochloride
Supplier: Matrix Scientific
053315-500MG
053315-2.5G
101903-146EA
364.62
USD
101903-146
101903-148
2-Amino-N-(2-methylphenyl)acetamide hydrochloride
2-Amino-N-(2-methylphenyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O |
MDL Number:
MFCD01733408 CAS Number: 1049692-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...