6-Chloro-N-(2-methylphenyl)-2-pyridinamine
Supplier: Matrix Scientific
053314-500MG
053314-2.5G
101903-142EA
364.62
USD
101903-142
101903-144
6-Chloro-N-(2-methylphenyl)-2-pyridinamine
6-Chloro-N-(2-methylphenyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₂H₁₁ClN₂ |
MDL Number:
MFCD13562707 CAS Number: 1220017-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...