N-(5-Bromo-2-pyridinyl)-N-(2-methylphenyl)amine

Supplier: Matrix Scientific

053313-500MG 053313-2.5G
101903-138EA 364.62 USD
101903-138 101903-140
N-(5-Bromo-2-pyridinyl)-N-(2-methylphenyl)amine
N-(5-Bromo-2-pyridinyl)-N-(2-methylphenyl)amine

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂
MDL Number: MFCD13562706
CAS Number: 1220030-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR