5-Bromo-3-methyl-N-(2-methylphenyl)-2-pyridinamine

Supplier: Matrix Scientific

053312-500MG 053312-2.5G
101903-134EA 364.62 USD
101903-134 101903-136
5-Bromo-3-methyl-N-(2-methylphenyl)-2-pyridinamine
5-Bromo-3-methyl-N-(2-methylphenyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂
MDL Number: MFCD13562705
CAS Number: 1220038-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR