N2-(2-Methylphenyl)-2,3-pyridinediamine

Supplier: Matrix Scientific

053310-500MG 053310-2.5G
101903-126EA 364.62 USD
101903-126 101903-128
N2-(2-Methylphenyl)-2,3-pyridinediamine
N2-(2-Methylphenyl)-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₂H₁₃N₃
MDL Number: MFCD11202141
CAS Number: 42048-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR