2-(2-Toluidino)isonicotinic acid
Supplier: Matrix Scientific
053308-500MG
053308-2.5G
101903-118EA
364.62
USD
101903-118
101903-120
2-(2-Toluidino)isonicotinic acid
2-(2-Toluidino)isonicotinic acid
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₂ |
MDL Number:
MFCD11132204 CAS Number: 1019324-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...