6-Chloro-N-(2-methylphenyl)-2-pyrazinamine
Supplier: Matrix Scientific
053307-500MG
053307-2.5G
101903-114EA
364.62
USD
101903-114
101903-116
6-Chloro-N-(2-methylphenyl)-2-pyrazinamine
6-Chloro-N-(2-methylphenyl)-2-pyrazinamine
Hazard: Irritant
Formula:
C₁₁H₁₀ClN₃ |
MDL Number:
MFCD13562704 CAS Number: 445264-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...