3-Chloro-N-(2-methylphenyl)-5-(trifluoromethyl)-2-pyridinamine
Supplier: Matrix Scientific
053306-500MG
053306-2.5G
101903-110EA
364.62
USD
101903-110
101903-112
3-Chloro-N-(2-methylphenyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(2-methylphenyl)-5-(trifluoromethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₃H₁₀ClF₃N₂ |
MDL Number:
MFCD13562703 CAS Number: 1220030-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...