6-Chloro-N-(2-methylphenyl)-4-pyrimidinamine

Supplier: Matrix Scientific

053305-500MG 053305-2.5G
101903-106EA 364.62 USD
101903-106 101903-108
6-Chloro-N-(2-methylphenyl)-4-pyrimidinamine
6-Chloro-N-(2-methylphenyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃
MDL Number: MFCD10037860
CAS Number: 936075-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR