5-Bromo-4-methyl-N-(2-methylphenyl)-2-pyridinamine
Supplier: Matrix Scientific
053304-500MG
053304-2.5G
101903-102EA
364.62
USD
101903-102
101903-104
5-Bromo-4-methyl-N-(2-methylphenyl)-2-pyridinamine
5-Bromo-4-methyl-N-(2-methylphenyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₃H₁₃BrN₂ |
MDL Number:
MFCD13562702 CAS Number: 1220038-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...