3-Nitro-4-(2-toluidino)benzoic acid

Supplier: Matrix Scientific

053303-500MG 053303-2.5G
101903-098EA 364.62 USD
101903-098 101903-100
3-Nitro-4-(2-toluidino)benzoic acid
3-Nitro-4-(2-toluidino)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₄
MDL Number: MFCD13354163
CAS Number: 860698-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR