3-Amino-4-chloro-N-(3-ethoxypropyl)benzenesulfonamide

Supplier: Matrix Scientific

053301-500MG 053301-2.5G
101903-090EA 364.62 USD
101903-090 101903-092
3-Amino-4-chloro-N-(3-ethoxypropyl)benzenesulfonamide
3-Amino-4-chloro-N-(3-ethoxypropyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O₃S
MW: 292.79 g/mol
MDL Number: MFCD11124989
CAS Number: 1040317-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR