N-(3-Ethoxypropyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053299-500MG 053299-2.5G
101903-082EA 364.62 USD
101903-082 101903-084
N-(3-Ethoxypropyl)-4-piperidinecarboxamide hydrochloride
N-(3-Ethoxypropyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O₂
MDL Number: MFCD13562699
CAS Number: 1220030-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR