N-(3-Ethoxypropyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053298-500MG 053298-2.5G
101903-078EA 364.62 USD
101903-078 101903-080
N-(3-Ethoxypropyl)-3-piperidinecarboxamide hydrochloride
N-(3-Ethoxypropyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O₂
MDL Number: MFCD13562698
CAS Number: 1220038-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR