2-Amino-N-(3-ethoxypropyl)-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053295-500MG
053295-2.5G
101903-066EA
364.62
USD
101903-066
101903-068
2-Amino-N-(3-ethoxypropyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(3-ethoxypropyl)-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₃ClN₂O₂ |
MDL Number:
MFCD13562695 CAS Number: 1236263-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...