2-Amino-N-(3-ethoxypropyl)-3-methylbutanamide hydrochloride
Supplier: Matrix Scientific
053293-500MG
053293-2.5G
101903-058EA
364.62
USD
101903-058
101903-060
2-Amino-N-(3-ethoxypropyl)-3-methylbutanamide hydrochloride
2-Amino-N-(3-ethoxypropyl)-3-methylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₃ClN₂O₂ |
MDL Number:
MFCD13562693 CAS Number: 1236261-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...