3-Amino-N-(3-ethoxypropyl)propanamide hydrochloride

Supplier: Matrix Scientific

053292-500MG 053292-2.5G
101903-054EA 364.62 USD
101903-054 101903-056
3-Amino-N-(3-ethoxypropyl)propanamide hydrochloride
3-Amino-N-(3-ethoxypropyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O₂
MDL Number: MFCD13562692
CAS Number: 1220035-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR