N-(3-Ethoxypropyl)-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

053291-500MG 053291-2.5G
101903-050EA 364.62 USD
101903-050 101903-052
N-(3-Ethoxypropyl)-2-(methylamino)acetamide hydrochloride
N-(3-Ethoxypropyl)-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O₂
MDL Number: MFCD13562691
CAS Number: 1220038-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR