2-Amino-N-(3-ethoxypropyl)propanamide hydrochloride
Supplier: Matrix Scientific
053290-500MG
053290-2.5G
101903-046EA
364.62
USD
101903-046
101903-048
2-Amino-N-(3-ethoxypropyl)propanamide hydrochloride
2-Amino-N-(3-ethoxypropyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₉ClN₂O₂ |
MDL Number:
MFCD13562690 CAS Number: 1246172-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...