6-Chloro-N-(3-ethoxypropyl)-2-pyridinamine
Supplier: Matrix Scientific
053288-500MG
053288-2.5G
101903-038EA
364.62
USD
101903-038
101903-040
6-Chloro-N-(3-ethoxypropyl)-2-pyridinamine
6-Chloro-N-(3-ethoxypropyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD13562688 CAS Number: 1220038-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...