N-(5-Bromo-2-pyridinyl)-N-(3-ethoxypropyl)amine

Supplier: Matrix Scientific

053287-500MG 053287-2.5G
101903-034EA 364.62 USD
101903-034 101903-036
N-(5-Bromo-2-pyridinyl)-N-(3-ethoxypropyl)amine
N-(5-Bromo-2-pyridinyl)-N-(3-ethoxypropyl)amine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂O
MDL Number: MFCD11116339
CAS Number: 1036555-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR