N-(5-Bromo-3-methyl-2-pyridinyl)-N-(3-ethoxypropyl)amine
Supplier: Matrix Scientific
053286-500MG
053286-2.5G
101903-030EA
364.62
USD
101903-030
101903-032
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(3-ethoxypropyl)amine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(3-ethoxypropyl)amine
Hazard: Irritant
Formula:
C₁₁H₁₇BrN₂O |
MDL Number:
MFCD13562687 CAS Number: 1220019-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...