2-Chloro-N-(3-ethoxypropyl)nicotinamide

Supplier: Matrix Scientific

053285-500MG 053285-2.5G
101903-026EA 364.62 USD
101903-026 101903-028
2-Chloro-N-(3-ethoxypropyl)nicotinamide
2-Chloro-N-(3-ethoxypropyl)nicotinamide

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD09816818
CAS Number: 1016718-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR