N-2-(3-Ethoxypropyl)-2,3-pyridinediamine
Supplier: Matrix Scientific
053284-500MG
053284-2.5G
101903-022EA
364.62
USD
101903-022
101903-024
N-2-(3-Ethoxypropyl)-2,3-pyridinediamine
N-2-(3-Ethoxypropyl)-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₁₀H₁₇N₃O |
MDL Number:
MFCD11187312 CAS Number: 1039967-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...