2-[(3-Ethoxypropyl)amino]nicotinic acid
Supplier: Matrix Scientific
053283-500MG
053283-2.5G
101903-018EA
364.62
USD
101903-018
101903-020
2-[(3-Ethoxypropyl)amino]nicotinic acid
2-[(3-Ethoxypropyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O₃ |
MDL Number:
MFCD11132182 CAS Number: 1019386-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...