2-[(3-Ethoxypropyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053282-500MG
053282-2.5G
101903-014EA
364.62
USD
101903-014
101903-016
2-[(3-Ethoxypropyl)amino]isonicotinic acid
2-[(3-Ethoxypropyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O₃ |
MDL Number:
MFCD11132179 CAS Number: 1019359-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...