6-Chloro-N-(3-ethoxypropyl)-2-pyrazinamine

Supplier: Matrix Scientific

053281-500MG 053281-2.5G
101903-010EA 364.62 USD
101903-010 101903-012
6-Chloro-N-(3-ethoxypropyl)-2-pyrazinamine
6-Chloro-N-(3-ethoxypropyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562686
CAS Number: 1220036-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR