3-Chloro-N-(3-ethoxypropyl)-5-(trifluoromethyl)-2-pyridinamine
Supplier: Matrix Scientific
053280-500MG
053280-2.5G
101903-006EA
364.62
USD
101903-006
101903-008
3-Chloro-N-(3-ethoxypropyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(3-ethoxypropyl)-5-(trifluoromethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₁H₁₄ClF₃N₂O |
MDL Number:
MFCD11125397 CAS Number: 1040309-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...