6-Chloro-N-(3-ethoxypropyl)-4-pyrimidinamine

Supplier: Matrix Scientific

053279-500MG 053279-2.5G
101903-002EA 364.62 USD
101903-002 101903-004
6-Chloro-N-(3-ethoxypropyl)-4-pyrimidinamine
6-Chloro-N-(3-ethoxypropyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562685
CAS Number: 1219976-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR