N-(5-Bromo-4-methyl-2-pyridinyl)-N-(3-ethoxypropyl)amine

Supplier: Matrix Scientific

053278-500MG 053278-2.5G
101902-998EA 364.62 USD
101902-998 101903-000
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(3-ethoxypropyl)amine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(3-ethoxypropyl)amine

Hazard: Irritant

Formula: C₁₁H₁₇BrN₂O
MDL Number: MFCD13562684
CAS Number: 1220036-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR