4-[(3-Ethoxypropyl)amino]-3-nitrobenzoic acid
Supplier: Matrix Scientific
053277-500MG
053277-2.5G
101902-994EA
364.62
USD
101902-994
101902-996
4-[(3-Ethoxypropyl)amino]-3-nitrobenzoic acid
4-[(3-Ethoxypropyl)amino]-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₅ |
MDL Number:
MFCD12550716 CAS Number: 1019452-53-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...