Ethyl-3-amino-4-[(3-ethoxypropyl)amino]benzoate

Supplier: Matrix Scientific

053276-500MG 053276-2.5G
101902-990EA 364.62 USD
101902-990 101902-992
Ethyl-3-amino-4-[(3-ethoxypropyl)amino]benzoate
Ethyl-3-amino-4-[(3-ethoxypropyl)amino]benzoate

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₃
MDL Number: MFCD13344458
CAS Number: 1220019-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR