3-Amino-4-chloro-N-[3-(dimethylamino)propyl]benzenesulfonamide hydrochloride
Supplier: Matrix Scientific
053275-500MG
053275-2.5G
101902-986EA
364.62
USD
101902-986
101902-988
3-Amino-4-chloro-N-[3-(dimethylamino)propyl]benzenesulfonamide hydrochloride
3-Amino-4-chloro-N-[3-(dimethylamino)propyl]benzenesulfonamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₉Cl₂N₃O₂S |
MDL Number:
MFCD13562683 CAS Number: 1219976-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...