N-[3-(Dimethylamino)propyl]-4-piperidinecarboxamide dihydrochloride
Supplier: Matrix Scientific
053273-500MG
053273-2.5G
101902-978EA
364.62
USD
101902-978
101902-980
N-[3-(Dimethylamino)propyl]-4-piperidinecarboxamide dihydrochloride
N-[3-(Dimethylamino)propyl]-4-piperidinecarboxamide dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₅Cl₂N₃O |
MDL Number:
MFCD13562681 CAS Number: 1220017-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...