N-[3-(Dimethylamino)propyl]-3-piperidinecarboxamide dihydrochloride

Supplier: Matrix Scientific

053272-500MG 053272-2.5G
101902-974EA 364.62 USD
101902-974 101902-976
N-[3-(Dimethylamino)propyl]-3-piperidinecarboxamide dihydrochloride
N-[3-(Dimethylamino)propyl]-3-piperidinecarboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅Cl₂N₃O
MDL Number: MFCD13562680
CAS Number: 1220036-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR