N-[3-(Dimethylamino)propyl]-2-piperidinecarboxamide dihydrochloride

Supplier: Matrix Scientific

053271-500MG 053271-2.5G
101902-970EA 364.62 USD
101902-970 101902-972
N-[3-(Dimethylamino)propyl]-2-piperidinecarboxamide dihydrochloride
N-[3-(Dimethylamino)propyl]-2-piperidinecarboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅Cl₂N₃O
MDL Number: MFCD13562679
CAS Number: 1236261-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR