N-[3-(Dimethylamino)propyl]-2-pyrrolidinecarboxamide dihydrochloride

Supplier: Matrix Scientific

053270-500MG 053270-2.5G
101902-966EA 364.62 USD
101902-966 101902-968
N-[3-(Dimethylamino)propyl]-2-pyrrolidinecarboxamide dihydrochloride
N-[3-(Dimethylamino)propyl]-2-pyrrolidinecarboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₃Cl₂N₃O
MDL Number: MFCD13562678
CAS Number: 1236267-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR