2-Amino-N-[3-(dimethylamino)propyl]-3-phenylpropanamide dihydrochloride
Supplier: Matrix Scientific
053269-500MG
053269-2.5G
101902-962EA
364.62
USD
101902-962
101902-964
2-Amino-N-[3-(dimethylamino)propyl]-3-phenylpropanamide dihydrochloride
2-Amino-N-[3-(dimethylamino)propyl]-3-phenylpropanamide dihydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₅Cl₂N₃O |
MDL Number:
MFCD13562677 CAS Number: 1236261-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...