2-Amino-N-[3-(dimethylamino)propyl]-2-methylpropanamide dihydrochloride
Supplier: Matrix Scientific
053268-500MG
053268-2.5G
101902-958EA
364.62
USD
101902-958
101902-960
2-Amino-N-[3-(dimethylamino)propyl]-2-methylpropanamide dihydrochloride
2-Amino-N-[3-(dimethylamino)propyl]-2-methylpropanamide dihydrochloride
Hazard: Irritant
Formula:
C₉H₂₃Cl₂N₃O |
MDL Number:
MFCD13562676 CAS Number: 1220037-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...