2-Amino-N-[3-(dimethylamino)propyl]-3-methylbutanamide dihydrochloride

Supplier: Matrix Scientific

053267-500MG 053267-2.5G
101902-954EA 364.62 USD
101902-954 101902-956
2-Amino-N-[3-(dimethylamino)propyl]-3-methylbutanamide dihydrochloride
2-Amino-N-[3-(dimethylamino)propyl]-3-methylbutanamide dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₅Cl₂N₃O
MDL Number: MFCD13562675
CAS Number: 1236263-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR