3-Amino-N-[3-(dimethylamino)propyl]propanamide dihydrochloride
Supplier: Matrix Scientific
053266-500MG
053266-2.5G
101902-950EA
364.62
USD
101902-950
101902-952
3-Amino-N-[3-(dimethylamino)propyl]propanamide dihydrochloride
3-Amino-N-[3-(dimethylamino)propyl]propanamide dihydrochloride
Hazard: Irritant
Formula:
C₈H₂₁Cl₂N₃O |
MDL Number:
MFCD13562674 CAS Number: 1220029-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...