N-[3-(Dimethylamino)propyl]-2-(methylamino)acetamide dihydrochloride
Supplier: Matrix Scientific
053265-500MG
053265-2.5G
101902-946EA
364.62
USD
101902-946
101902-948
N-[3-(Dimethylamino)propyl]-2-(methylamino)acetamide dihydrochloride
N-[3-(Dimethylamino)propyl]-2-(methylamino)acetamide dihydrochloride
Hazard: Irritant
Formula:
C₈H₂₁Cl₂N₃O MW: 246.18 g/mol |
MDL Number:
MFCD13562673 CAS Number: 1220036-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...