2-Amino-N-[3-(dimethylamino)propyl]propanamide dihydrochloride

Supplier: Matrix Scientific

053264-500MG 053264-2.5G
101902-942EA 364.62 USD
101902-942 101902-944
2-Amino-N-[3-(dimethylamino)propyl]propanamide dihydrochloride
2-Amino-N-[3-(dimethylamino)propyl]propanamide dihydrochloride

Hazard: Irritant

Formula: C₈H₂₁Cl₂N₃O
MDL Number: MFCD13562672
CAS Number: 1236255-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR