2-Amino-N-[3-(dimethylamino)propyl]acetamide dihydrochloride

Supplier: Matrix Scientific

053263-500MG 053263-2.5G
101902-938EA 364.62 USD
101902-938 101902-940
2-Amino-N-[3-(dimethylamino)propyl]acetamide dihydrochloride
2-Amino-N-[3-(dimethylamino)propyl]acetamide dihydrochloride

Hazard: Irritant

Formula: C₇H₁₉Cl₂N₃O
MDL Number: MFCD13562671
CAS Number: 1220029-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR