N1-(5-Bromo-2-pyridinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Supplier: Matrix Scientific

053261-500MG 053261-2.5G
101902-930EA 364.62 USD
101902-930 101902-932
N1-(5-Bromo-2-pyridinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride
N1-(5-Bromo-2-pyridinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇BrClN₃
MDL Number: MFCD13562669
CAS Number: 1220029-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR