N1-(5-Bromo-3-methyl-2-pyridinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Supplier: Matrix Scientific

053260-500MG 053260-2.5G
101902-926EA 364.62 USD
101902-926 101902-928
N1-(5-Bromo-3-methyl-2-pyridinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride
N1-(5-Bromo-3-methyl-2-pyridinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₉BrClN₃
MDL Number: MFCD13562668
CAS Number: 1220017-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR